missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals L-Tyrosine, 99%

CAS: 60-18-4 | C9H11NO3 | 181.19 g/mol

Supplier:  Thermo Scientific Chemicals A111410B

 View more versions of this product

Catalog No. AAA111410B


Only null left

Description

Description

L-Tyrosine is used in pharmaceuticals, dietary supplements and food additives. It is a precursor to alkaloids such as morphine, neurotransmitters, epinephrine, p-coumaric acid, thyroxine, pigment melanin and catcholamines. It plays a vital role in photosynthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
L-Tyrosine is used in pharmaceuticals, dietary supplements and food additives. It is a precursor to alkaloids such as morphine, neurotransmitters, epinephrine, p-coumaric acid, thyroxine, pigment melanin and catcholamines. It plays a vital role in photosynthesis.

Solubility
Soluble in hydrochloric acid. Slightly soluble in water.

Notes
Incompatible with strong oxidizing agents and strong reducing agents.
Specifications

Specifications

>300°C
60-18-4
C9H11NO3
392441
l-tyrosine, tyrosine, s-tyrosine, p-tyrosine, h-tyr-oh, l-p-tyrosine, 2s-2-amino-3-4-hydroxyphenyl propanoic acid, 4-hydroxy-l-phenylalanine, l---tyrosine, tyrosine, l
OUYCCCASQSFEME-SVGMAFHSNA-N
N[C@@H](CC1=CC=C(O)C=C1)C(O)=O
181.19
6057
−11° (c=4 in 1N HCl)
99%
L-Tyrosine
99%
MFCD00002606
14,9839
Soluble in hydrochloric acid. Slightly soluble in water.
1.46
2-amino-3-(4-hydroxyphenyl)propanoic acid
1000 g
CHEBI:17895
181.19

Chemical Identifiers

60-18-4
181.19
OUYCCCASQSFEME-SVGMAFHSNA-N
6057
N[C@@H](CC1=CC=C(O)C=C1)C(O)=O
C9H11NO3
MFCD00002606
l-tyrosine, tyrosine, s-tyrosine, p-tyrosine, h-tyr-oh, l-p-tyrosine, 2s-2-amino-3-4-hydroxyphenyl propanoic acid, 4-hydroxy-l-phenylalanine, l---tyrosine, tyrosine, l
CHEBI:17895
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

EINECSNumber : 200-460-4

RTECSNumber : YP2275600

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only