missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals L-Tyrosine, 99%

Catalog No. p-7081223
Change view
Click to view available options
Quantity:
50 g
100 g
250 g
1000 g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1114118 50 g
AAA1114122 100 g
AAA1114130 250 g
AAA111410B 1000 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. AAA1114118 Supplier Thermo Scientific Chemicals Supplier No. A1114118

CAS: 60-18-4 | C9H11NO3 | 181.19 g/mol

L-Tyrosine is used in pharmaceuticals, dietary supplements and food additives. It is a precursor to alkaloids such as morphine, neurotransmitters, epinephrine, p-coumaric acid, thyroxine, pigment melanin and catcholamines. It plays a vital role in photosynthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
L-Tyrosine is used in pharmaceuticals, dietary supplements and food additives. It is a precursor to alkaloids such as morphine, neurotransmitters, epinephrine, p-coumaric acid, thyroxine, pigment melanin and catcholamines. It plays a vital role in photosynthesis.

Solubility
Soluble in hydrochloric acid. Slightly soluble in water.

Notes
Incompatible with strong oxidizing agents and strong reducing agents.

Chemical Identifiers

CAS 60-18-4
Molecular Formula C9H11NO3
Molecular Weight (g/mol) 181.19
MDL Number MFCD00002606
InChI Key OUYCCCASQSFEME-SVGMAFHSNA-N
Synonym l-tyrosine, tyrosine, s-tyrosine, p-tyrosine, h-tyr-oh, l-p-tyrosine, 2s-2-amino-3-4-hydroxyphenyl propanoic acid, 4-hydroxy-l-phenylalanine, l---tyrosine, tyrosine, l
PubChem CID 6057
ChEBI CHEBI:17895
SMILES N[C@@H](CC1=CC=C(O)C=C1)C(O)=O

Specifications

Melting Point >300°C
Assay Percent Range 99%
Beilstein 392441
Merck Index 14,9839
Solubility Information Soluble in hydrochloric acid. Slightly soluble in water.
Density 1.46 g/mL
IUPAC Name (2S)-2-amino-3-(4-hydroxyphenyl)propanoic acid
Quantity 50 g
Formula Weight 181.19
Percent Purity 99%
Chemical Name or Material L-Tyrosine
Show More Show Less
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 200-460-4
RTECSNumber YP2275600
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only