missing translation for 'onlineSavingsMsg'
Learn More

L-(-)-Tryptophanol 97.0+%, TCI America™

Catalog No. T15371G
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T15371G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. T15371G Supplier TCI America Supplier No. T15371G

Chemical Identifiers

CAS 2899-29-8
Molecular Formula C11H14N2O
Molecular Weight (g/mol) 190.25
MDL Number MFCD00037970
InChI Key UDQCRUSSQAXPJY-UHFFFAOYNA-N
Synonym l-tryptophanol, h-tryptophanol, l---tryptophanol, tryptophanol, s-2-amino-3-1h-indol-3-yl propan-1-ol, 2s-2-amino-3-1h-indol-3-yl propan-1-ol, s-2-amino-3-3-indolyl-1-propanol, s---2-amino-3-3-indolyl propanol, tryptophanol, l, 2-amino-3-1h-indol-3-yl-propan-1-ol
PubChem CID 6951149
IUPAC Name 2-amino-3-(1H-indol-3-yl)propan-1-ol
SMILES NC(CO)CC1=CNC2=CC=CC=C12

Specifications

Quantity 1 g
Formula Weight 190.25
Percent Purity ≥97.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material L-(-)-Tryptophanol