accessibility menu, dialog, popup

UserName

NH(S)NH2OH

CAS RN 2899-29-8

IUPAC Name: 2-amino-3-(1H-indol-3-yl)propan-1-ol
Synonyms: 2-amino-3-(1H-indol-3-yl)propan-1-ol
Molecular Weight (g/mol): 190.25
Molecular Formula: C11H14N2O
InChi Key: UDQCRUSSQAXPJY-UHFFFAOYNA-N
SMILES: NC(CO)CC1=CNC2=CC=CC=C12
Percent Purity 
  • (1)
  • (2)
Quantity 
  • (2)
  • (1)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results