missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals L-Tryptophanol, 97%

Catalog No. AAH6250603
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6250603 1 g
AAH6250606 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH6250603 Supplier Thermo Scientific Chemicals Supplier No. H6250603

CAS: 2899-29-8 | C11H14N2O | 190.25 g/mol

L-Tryptophanol is an amino acid used in the construction of peptides and proteins.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
L-Tryptophanol is an amino acid used in the construction of peptides and proteins.

Solubility
Soluble in water 11.4 g/L (25°C).

Notes
Store in a cool, dry place, in a well sealed container. Store away from oxidizing agents.

Chemical Identifiers

CAS 2899-29-8
Molecular Formula C11H14N2O
Molecular Weight (g/mol) 190.25
MDL Number MFCD00037970
InChI Key UDQCRUSSQAXPJY-UHFFFAOYNA-N
Synonym l-tryptophanol, h-tryptophanol, l---tryptophanol, tryptophanol, s-2-amino-3-1h-indol-3-yl propan-1-ol, 2s-2-amino-3-1h-indol-3-yl propan-1-ol, s-2-amino-3-3-indolyl-1-propanol, s---2-amino-3-3-indolyl propanol, tryptophanol, l, 2-amino-3-1h-indol-3-yl-propan-1-ol
PubChem CID 6951149
IUPAC Name 2-amino-3-(1H-indol-3-yl)propan-1-ol
SMILES NC(CO)CC1=CNC2=CC=CC=C12

Specifications

Color Brown
Solubility Information Soluble in water 11.4g/L (25°C).
Quantity 1 g
Formula Weight 190.24
Percent Purity 97%
Physical Form Viscous Liquid
Chemical Name or Material L-Tryptophanol
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only