missing translation for 'onlineSavingsMsg'
Learn More
Learn More
L-Tryptophanamide Hydrochloride 98.0+%, TCI America™
$128.98 - $512.10
Chemical Identifiers
| CAS | 5022-65-1 |
|---|---|
| Molecular Formula | C11H14ClN3O |
| Molecular Weight (g/mol) | 239.70 |
| MDL Number | MFCD00054315 |
| InChI Key | WOBDANBSEWOYKN-FVGYRXGTSA-N |
| Synonym | l-tryptophanamide hydrochloride, h-trp-nh2.hcl, s-2-amino-3-1h-indol-3-yl propanamide hydrochloride, h-trp-nh2 hcl, l-tryptophane amide hydrochloride, 2s-2-amino-3-1h-indol-3-yl propanamide hydrochloride, l---tryptophan methyl ester hydrochloride, h-trp-nh hcl, h-trp-nh2 . hcl |
| PubChem CID | 6452180 |
| IUPAC Name | (2S)-2-amino-3-(1H-indol-3-yl)propanamide hydrochloride |
| SMILES | Cl.N[C@@H](CC1=CNC2=CC=CC=C12)C(N)=O |
Chemical Identifiers
| 5022-65-1 | |
| 239.70 | |
| WOBDANBSEWOYKN-FVGYRXGTSA-N | |
| 6452180 | |
| Cl.N[C@@H](CC1=CNC2=CC=CC=C12)C(N)=O |
| C11H14ClN3O | |
| MFCD00054315 | |
| l-tryptophanamide hydrochloride, h-trp-nh2.hcl, s-2-amino-3-1h-indol-3-yl propanamide hydrochloride, h-trp-nh2 hcl, l-tryptophane amide hydrochloride, 2s-2-amino-3-1h-indol-3-yl propanamide hydrochloride, l---tryptophan methyl ester hydrochloride, h-trp-nh hcl, h-trp-nh2 . hcl | |
| (2S)-2-amino-3-(1H-indol-3-yl)propanamide hydrochloride |
Specifications
| 5022-65-1 | |
| White-Yellow | |
| MFCD00054315 | |
| l-tryptophanamide hydrochloride, h-trp-nh2.hcl, s-2-amino-3-1h-indol-3-yl propanamide hydrochloride, h-trp-nh2 hcl, l-tryptophane amide hydrochloride, 2s-2-amino-3-1h-indol-3-yl propanamide hydrochloride, l---tryptophan methyl ester hydrochloride, h-trp-nh hcl, h-trp-nh2 . hcl | |
| Cl.N[C@@H](CC1=CNC2=CC=CC=C12)C(N)=O | |
| 239.70 | |
| 239.70 | |
| Crystalline Powder |
| 250°C | |
| C11H14ClN3O | |
| 1 g | |
| WOBDANBSEWOYKN-FVGYRXGTSA-N | |
| (2S)-2-amino-3-(1H-indol-3-yl)propanamide hydrochloride | |
| 6452180 | |
| ≥98.0% (HPLC,N) | |
| L-Tryptophanamide Hydrochloride |
Safety and Handling
TSCA : No