missing translation for 'onlineSavingsMsg'
Learn More

L-Tryptophanamide Hydrochloride 98.0+%, TCI America™

Catalog No. T23121G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T23121G 1 g
T23125G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T23121G Supplier TCI America Supplier No. T23121G
Only null left

Chemical Identifiers

CAS 5022-65-1
Molecular Formula C11H14ClN3O
Molecular Weight (g/mol) 239.70
MDL Number MFCD00054315
InChI Key WOBDANBSEWOYKN-FVGYRXGTSA-N
Synonym l-tryptophanamide hydrochloride, h-trp-nh2.hcl, s-2-amino-3-1h-indol-3-yl propanamide hydrochloride, h-trp-nh2 hcl, l-tryptophane amide hydrochloride, 2s-2-amino-3-1h-indol-3-yl propanamide hydrochloride, l---tryptophan methyl ester hydrochloride, h-trp-nh hcl, h-trp-nh2 . hcl
PubChem CID 6452180
IUPAC Name (2S)-2-amino-3-(1H-indol-3-yl)propanamide hydrochloride
SMILES Cl.N[C@@H](CC1=CNC2=CC=CC=C12)C(N)=O

Specifications

Melting Point 250°C
Color White-Yellow
Quantity 1 g
Formula Weight 239.70
Percent Purity ≥98.0% (HPLC,N)
Physical Form Crystalline Powder
Chemical Name or Material L-Tryptophanamide Hydrochloride