missing translation for 'onlineSavingsMsg'
Learn More
Learn More
L-Tryptophanamide hydrochloride, 95%
CAS: 5022-65-1 | C11H14ClN3O | 239.70 g/mol
Chemical Identifiers
| CAS | 5022-65-1 |
|---|---|
| Molecular Formula | C11H14ClN3O |
| Molecular Weight (g/mol) | 239.70 |
| MDL Number | MFCD00054315 |
| InChI Key | WOBDANBSEWOYKN-FVGYRXGTSA-N |
| Synonym | l-tryptophanamide hydrochloride, h-trp-nh2.hcl, s-2-amino-3-1h-indol-3-yl propanamide hydrochloride, h-trp-nh2 hcl, l-tryptophane amide hydrochloride, 2s-2-amino-3-1h-indol-3-yl propanamide hydrochloride, l---tryptophan methyl ester hydrochloride, h-trp-nh hcl, h-trp-nh2 . hcl |
| PubChem CID | 6452180 |
| SMILES | Cl.N[C@@H](CC1=CNC2=CC=CC=C12)C(N)=O |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|
AAH6340503
|
N/A | 1 g |
N/A
|
|
|||||
|
AAH6340506
|
N/A | 5 g |
N/A
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| C11H14ClN3O | |
| MFCD00054315 | |
| l-tryptophanamide hydrochloride, h-trp-nh2.hcl, s-2-amino-3-1h-indol-3-yl propanamide hydrochloride, h-trp-nh2 hcl, l-tryptophane amide hydrochloride, 2s-2-amino-3-1h-indol-3-yl propanamide hydrochloride, l---tryptophan methyl ester hydrochloride, h-trp-nh hcl, h-trp-nh2 . hcl | |
| Cl.N[C@@H](CC1=CNC2=CC=CC=C12)C(N)=O |
Specifications
| 5022-65-1 | |
| MFCD00054315 | |
| WOBDANBSEWOYKN-FVGYRXGTSA-N | |
| hydrogen (2S)-2-amino-3-(1H-indol-3-yl)propanamide chloride | |
| 6452180 | |
| 95% | |
| L-Tryptophanamide hydrochloride |
| C11H14ClN3O | |
| l-tryptophanamide hydrochloride, h-trp-nh2.hcl, s-2-amino-3-1h-indol-3-yl propanamide hydrochloride, h-trp-nh2 hcl, l-tryptophane amide hydrochloride, 2s-2-amino-3-1h-indol-3-yl propanamide hydrochloride, l---tryptophan methyl ester hydrochloride, h-trp-nh hcl, h-trp-nh2 . hcl | |
| Cl.N[C@@H](CC1=CNC2=CC=CC=C12)C(N)=O | |
| 239.70 | |
| 239.7 | |
| Solid |
Safety and Handling
EINECSNumber : 225-708-9
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only