missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Specifications
| L-Tryptophan | |
| 282°C | |
| C11H12N2O2 | |
| 25 g | |
| QIVBCDIJIAJPQS-VIFPVBQESA-N | |
| (2S)-2-amino-3-(1H-indol-3-yl)propanoic acid | |
| 6305 | |
| 204.23 | |
| Crystalline Powder |
| 73-22-3 | |
| White | |
| MFCD00064340 | |
| l-tryptophan, tryptophan, l-tryptophane, s-tryptophan, tryptophane, h-trp-oh, optimax, trofan, tryptacin, ardeytropin | |
| N[C@@H](CC1=CNC2=CC=CC=C12)C(O)=O | |
| 204.23 | |
| CHEBI:16828 | |
| ≥98.5% (HPLC,T) |
Chemical Identifiers
| 73-22-3 | |
| 204.23 | |
| QIVBCDIJIAJPQS-VIFPVBQESA-N | |
| 6305 | |
| (2S)-2-amino-3-(1H-indol-3-yl)propanoic acid |
| C11H12N2O2 | |
| MFCD00064340 | |
| l-tryptophan, tryptophan, l-tryptophane, s-tryptophan, tryptophane, h-trp-oh, optimax, trofan, tryptacin, ardeytropin | |
| CHEBI:16828 | |
| N[C@@H](CC1=CNC2=CC=CC=C12)C(O)=O |
Safety and Handling
EINECSNumber : (9)-0869
RTECSNumber : YN6130000
TSCA : Yes