Learn More
Thermo Scientific Chemicals L-Tryptophan, Cell Culture Reagent
CAS: 73-22-3 | C11H12N2O2 | 204.23 g/mol
Supplier: Thermo Scientific Chemicals J6250822
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
| 280°C to 285°C | |
| 73-22-3 | |
| C11H12N2O2 | |
| 86197 | |
| l-tryptophan, tryptophan, l-tryptophane, s-tryptophan, tryptophane, h-trp-oh, optimax, trofan, tryptacin, ardeytropin | |
| N[C@@H](CC1=CNC2=CC=CC=C12)C(O)=O | |
| 100 g | |
| 6305 | |
| 204.23 | |
| Powder |
| L-Tryptophan | |
| White | |
| MFCD00064340 | |
| 14,9797 | |
| QIVBCDIJIAJPQS-VIFPVBQESA-N | |
| (2S)-2-amino-3-(1H-indol-3-yl)propanoic acid | |
| 204.23 | |
| CHEBI:16828 | |
| Cell Culture Reagent |
Chemical Identifiers
| 73-22-3 | |
| 204.23 | |
| QIVBCDIJIAJPQS-VIFPVBQESA-N | |
| 6305 | |
| (2S)-2-amino-3-(1H-indol-3-yl)propanoic acid |
| C11H12N2O2 | |
| MFCD00064340 | |
| l-tryptophan, tryptophan, l-tryptophane, s-tryptophan, tryptophane, h-trp-oh, optimax, trofan, tryptacin, ardeytropin | |
| CHEBI:16828 | |
| N[C@@H](CC1=CNC2=CC=CC=C12)C(O)=O |
Safety and Handling
EINECSNumber : 200-795-6
RTECSNumber : YN6130000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only