missing translation for 'onlineSavingsMsg'
Learn More

L-Thyroxine, 98.5%, MP Biomedicals™

Catalog No. ICN15214501
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN15214501 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN15214501 Supplier MP Biomedicals Inc Supplier No. 0215214501

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

A thyroid hormone used in metabolic research to study hormonal conversion and protein synthesis regulation.

Chemical Identifiers

CAS 51-48-9
Molecular Formula C15H11I4NO4
Molecular Weight (g/mol) 776.87
MDL Number MFCD00002595
InChI Key XUIIKFGFIJCVMT-UHFFFAOYNA-N
Synonym l-thyroxine, levothyroxine, thyroxine, thyroxin, synthroid, tetraiodothyronine, thyrax, levothyroxin, thyratabs, thyreoideum
PubChem CID 5819
ChEBI CHEBI:18332
IUPAC Name 2-amino-3-[4-(4-hydroxy-3,5-diiodophenoxy)-3,5-diiodophenyl]propanoic acid
SMILES NC(CC1=CC(I)=C(OC2=CC(I)=C(O)C(I)=C2)C(I)=C1)C(O)=O

Specifications

Color White to Yellow
Assay Percent Range 0.985
Quantity 1 g
Formula Weight 776.9
Grade Not Available
Physical Form Crystalline Powder
Chemical Name or Material L-Thyroxine