accessibility menu, dialog, popup

UserName

H2NIOIHOIIHOO

CAS RN 51-48-9

IUPAC Name: 2-amino-3-[4-(4-hydroxy-3,5-diiodophenoxy)-3,5-diiodophenyl]propanoic acid
Synonyms: thyroxine dextrothyroxinum natricum dekstrotyroksiininatrium lisolipin dextrotyroxinnatrium D-thyroxine sodium Thx dextrotiroxina sódica eulipos dynothel
Molecular Weight (g/mol): 776.87
Molecular Formula: C15H11I4NO4
InChi Key: XUIIKFGFIJCVMT-UHFFFAOYNA-N
SMILES: NC(CC1=CC(I)=C(OC2=CC(I)=C(O)C(I)=C2)C(I)=C1)C(O)=O
Physical Form 
  • (3)
Quantity 
  • (2)
  • (1)
Grade 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (3)
  • (2)
  • (1)
  • (1)