missing translation for 'onlineSavingsMsg'
Learn More

L-Threonine methyl ester hydrochloride, 98%

Catalog No. AAH6211803
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6211803 1 g
AAH6211806 5 g
AAH6211814 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAH6211803 Supplier Thermo Scientific Chemicals Supplier No. H6211803
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 39994-75-7 | C5H12ClNO3 | 169.61 g/mol

L-Threonine methyl ester hydrochloride is used as a feed and food additive.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
L-Threonine methyl ester hydrochloride is used as a feed and food additive.

Solubility
Soluble in water.

Notes
Moisture Sensitive and Hygroscopic. Keep container tightly closed. Store away from water/moisture.

Chemical Identifiers

CAS 39994-75-7
Molecular Formula C5H12ClNO3
Molecular Weight (g/mol) 169.61
MDL Number MFCD00037677
InChI Key OZSJLLVVZFTDEY-HJXLNUONSA-N
Synonym l-threonine methyl ester hydrochloride, methyl l-threoninate hydrochloride, h-thr-ome.hcl, 2s,3r-methyl 2-amino-3-hydroxybutanoate hydrochloride, l-threonine, methyl ester, hydrochloride, l-threonine methyl ester hcl, threonine methyl ester hydrochloride, methyl 2s,3r-2-amino-3-hydroxybutanoate hydrochloride, methyl l-threoninate hcl, h-thr-ome?cl
PubChem CID 2734893
IUPAC Name (2S,3R)-3-hydroxy-1-methoxy-1-oxobutan-2-aminium
SMILES Cl.COC(=O)[C@@H](N)[C@@H](C)O

Specifications

Beilstein 3562717
Solubility Information Soluble in water.
Quantity 1 g
Formula Weight 169.61
Percent Purity 98%
Sensitivity Hygroscopic; Moisture sensitive
Chemical Name or Material L-Threonine methyl ester hydrochloride
EINECSNumber 254-738-5
TSCA No
Recommended Storage Store at -20°C; Store under Nitrogen

RUO – Research Use Only