missing translation for 'onlineSavingsMsg'
Learn More
Learn More
L-Threonine, Crystal, FCC, 98.5-101.5%, Spectrum™ Chemical
T1057, 72-19-5, C4H9NO3
Supplier: Spectrum Chemical Mfg Cor T1057100GM
Description
Spectrum™ Chemical L-Threonine, Crystal, FCC - The FCC grade meets the requirements of the Food Chemical Codex indicates and is suitable for all food, beverage and nutritional supplement applications. Spectrum Chemical offers over 300 Food grade chemical ingredients packaged in laboratory size bottles to production drum quantities and are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.Specifications
| 72-19-5 | |
| 0.1% | |
| Poly Bottle | |
| 100 g | |
| -26.5° to -29.0° | |
| 2-amino-3-hydroxybutanoic acid | |
| 98.5 to 101.5% |
| 100% | |
| 0.2% | |
| C4H9NO3 | |
| AYFVYJQAPQTCCC-UHFFFAOYNA-N | |
| CC(O)C(N)C(O)=O | |
| 119.12 | |
| FCC |
Chemical Identifiers
| 72-19-5 | |
| 119.12 | |
| 2-amino-3-hydroxybutanoic acid |
| C4H9NO3 | |
| AYFVYJQAPQTCCC-UHFFFAOYNA-N | |
| CC(O)C(N)C(O)=O |