missing translation for 'onlineSavingsMsg'
Learn More

L-Threonine, Crystal, FCC, 98.5-101.5%, Spectrum™ Chemical

Catalog No. p-7157852
Change view
Click to view available options
Quantity:
25 g
100 g
1 kg
Packaging:
Poly Bottle
Poly Pail
Catalog No. Quantity Packaging
18603982 25 g Poly Bottle
18603983 100 g Poly Bottle
18603984 1 kg Poly Pail
3 options

Catalog No. 18603982

Supplier: Spectrum Chemical Mfg Cor T105725GM

Only null left

T1057, 72-19-5, C4H9NO3

Spectrum™ Chemical L-Threonine, Crystal, FCC - The FCC grade meets the requirements of the Food Chemical Codex indicates and is suitable for all food, beverage and nutritional supplement applications. Spectrum Chemical offers over 300 Food grade chemical ingredients packaged in laboratory size bottles to production drum quantities and are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 72-19-5
Molecular Formula C4H9NO3
Molecular Weight (g/mol) 119.12
InChI Key AYFVYJQAPQTCCC-UHFFFAOYNA-N
IUPAC Name 2-amino-3-hydroxybutanoic acid
SMILES CC(O)C(N)C(O)=O

Specifications

CAS Max % 100%
Ignition Residue 0.1%
Loss on Drying 0.2%
Packaging Poly Bottle
Quantity 25 g
Optical Rotation -26.5° to -29.0°
Percent Purity 98.5 to 101.5%
Grade FCC