missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals L-Threonine, Cell Culture Reagent

CAS: 72-19-5 | C4H9NO3 | 119.12 g/mol

Supplier:  Thermo Scientific Chemicals J63709A1

 View more versions of this product

Catalog No. AAJ63709A1


Only null left
Explore available promotions

Description

Description

Threonine is used to treat various nervous system disorders including spinal spasticity, multiple sclerosis, familial spastic paraparesis, and amyotrophic lateral sclerosis (ALS, Lou Gehrig's disease). Useful with intestinal disorders and indigestion, threonine has also been used to alleviate anxiety and mild depression. . L-threonine has been used clinically with the aim of increasing glycine concentrations in the cerebral spinal fluid of patients with spasticity.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Solubility
Soluble in water 50 mg/mL.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
Specifications

Specifications

256°C (decomposition)
72-19-5
C4H9NO3
1721646
l-threonine, threonine, threonin, h-thr-oh, 2s,3r-2-amino-3-hydroxybutanoic acid, s-threonine, l---threonine, 2-amino-3-hydroxybutyric acid, threonine van, threoninum latin
AYFVYJQAPQTCCC-UHFFFAOYNA-N
(2S,3R)-2-amino-3-hydroxybutanoic acid
119.12
CHEBI:16857
Cell Culture Reagent
L-Threonine
White
MFCD00064270
14,9380
Soluble in water 50mg/mL.
CC(O)C(N)C(O)=O
1 kg
6288
119.12
Powder

Chemical Identifiers

72-19-5
119.12
AYFVYJQAPQTCCC-UHFFFAOYNA-N
6288
(2S,3R)-2-amino-3-hydroxybutanoic acid
C4H9NO3
MFCD00064270
l-threonine, threonine, threonin, h-thr-oh, 2s,3r-2-amino-3-hydroxybutanoic acid, s-threonine, l---threonine, 2-amino-3-hydroxybutyric acid, threonine van, threoninum latin
CHEBI:16857
CC(O)C(N)C(O)=O
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

EINECSNumber : 200-774-1

RTECSNumber : XO8590000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

Promotions

Promotions

RUO – Research Use Only