missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals L-Threonine, Cell Culture Reagent

Catalog No. p-7042775
Change view
Click to view available options
Quantity:
250 g
1 kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ6370930 250 g
AAJ63709A1 1 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAJ6370930 Supplier Thermo Scientific Chemicals Supplier No. J6370930
Only null left

CAS: 72-19-5 | C4H9NO3 | 119.12 g/mol

Threonine is used to treat various nervous system disorders including spinal spasticity, multiple sclerosis, familial spastic paraparesis, and amyotrophic lateral sclerosis (ALS, Lou Gehrig's disease). Useful with intestinal disorders and indigestion, threonine has also been used to alleviate anxiety and mild depression. . L-threonine has been used clinically with the aim of increasing glycine concentrations in the cerebral spinal fluid of patients with spasticity.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Solubility
Soluble in water 50 mg/mL.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.

Chemical Identifiers

CAS 72-19-5
Molecular Formula C4H9NO3
Molecular Weight (g/mol) 119.12
MDL Number MFCD00064270
InChI Key AYFVYJQAPQTCCC-UHFFFAOYNA-N
Synonym l-threonine, threonine, threonin, h-thr-oh, 2s,3r-2-amino-3-hydroxybutanoic acid, s-threonine, l---threonine, 2-amino-3-hydroxybutyric acid, threonine van, threoninum latin
PubChem CID 6288
ChEBI CHEBI:16857
IUPAC Name (2S,3R)-2-amino-3-hydroxybutanoic acid
SMILES CC(O)C(N)C(O)=O

Specifications

Melting Point 256°C (decomposition)
Color White
Beilstein 1721646
Merck Index 14,9380
Solubility Information Soluble in water 50mg/mL.
Quantity 250 g
Formula Weight 119.12
Grade Cell Culture Reagent
Physical Form Powder
Chemical Name or Material L-Threonine
EINECSNumber 200-774-1
RTECSNumber XO8590000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only