missing translation for 'onlineSavingsMsg'
Learn More
Learn More
L-Threonine Allo-free base, 990%, MP Biomedicals™
L-threonine is classified as an essential amino acid. Synthesized from homoserine phosphate by threonine synthase or from acetaldehyde and glycine by serine hydroxymethyltransferase
Supplier: MP Biomedicals Inc 0210305390
| Quantity | 500 g |
|---|
Chemical Identifiers
| C4H9NO3 | |
| MFCD00064270 | |
| l-threonine, threonine, threonin, h-thr-oh, 2s,3r-2-amino-3-hydroxybutanoic acid, s-threonine, l---threonine, 2-amino-3-hydroxybutyric acid, threonine van, threoninum latin | |
| CHEBI:16857 | |
| CC(O)C(N)C(O)=O |
Specifications
| L-Threonine | |
| 72-19-5 | |
| 99% | |
| MFCD00064270 | |
| AYFVYJQAPQTCCC-UHFFFAOYNA-N | |
| 2-amino-3-hydroxybutanoic acid | |
| 119.12 | |
| CHEBI:16857 | |
| 99.5% |
| Allo Free | |
| White | |
| C4H9NO3 | |
| l-threonine, threonine, threonin, h-thr-oh, 2s,3r-2-amino-3-hydroxybutanoic acid, s-threonine, l---threonine, 2-amino-3-hydroxybutyric acid, threonine van, threoninum latin | |
| CC(O)C(N)C(O)=O | |
| 500 g | |
| 6288 | |
| 119.1 | |
| Crystalline Powder |
Safety and Handling
EINECSNumber : 200-774-1
Recommended Storage : Store at Room temperature (15 to 30°C).