missing translation for 'onlineSavingsMsg'
Learn More

L-Threonine Allo-free base, 990%, MP Biomedicals™

Catalog No. p-4404551
Change view
Click to view available options
Quantity:
10 g
25 g
100 g
500 g
1 kg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN10305310 10 g
ICN10305325 25 g
ICN10305380 100 g
ICN10305390 500 g
ICN10305391 1 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. ICN10305310 Supplier MP Biomedicals Inc Supplier No. 0210305310
Only null left

L-threonine is classified as an essential amino acid. Synthesized from homoserine phosphate by threonine synthase or from acetaldehyde and glycine by serine hydroxymethyltransferase

Chemical Identifiers

CAS 72-19-5
Molecular Formula C4H9NO3
Molecular Weight (g/mol) 119.12
MDL Number MFCD00064270
InChI Key AYFVYJQAPQTCCC-UHFFFAOYNA-N
Synonym l-threonine, threonine, threonin, h-thr-oh, 2s,3r-2-amino-3-hydroxybutanoic acid, s-threonine, l---threonine, 2-amino-3-hydroxybutyric acid, threonine van, threoninum latin
PubChem CID 6288
ChEBI CHEBI:16857
IUPAC Name 2-amino-3-hydroxybutanoic acid
SMILES CC(O)C(N)C(O)=O

Specifications

Color White
Assay Percent Range 99%
Quantity 10 g
Formula Weight 119.1
Percent Purity 99.5%
Physical Form Crystalline Powder
Chemical Name or Material L-Threonine, Allo Free
EINECSNumber 200-774-1
Recommended Storage Store at Room temperature (15 to 30°C).