missing translation for 'onlineSavingsMsg'
Learn More
Learn More
L-Threonine, ≥98.5%, Ultrapure
CAS: 72-19-5 | C4H9NO3 | 119.12 g/mol
$216.78 - $1,541.92
Chemical Identifiers
| CAS | 72-19-5 |
|---|---|
| Molecular Formula | C4H9NO3 |
| Molecular Weight (g/mol) | 119.12 |
| MDL Number | MFCD00064270 |
| InChI Key | AYFVYJQAPQTCCC-UHFFFAOYNA-N |
| Synonym | l-threonine, threonine, threonin, h-thr-oh, 2 s,3 r-2-amino-3-hydroxybutanoic acid, s-threonine, l---threonine, 2-amino-3-hydroxybutyric acid, threonine van, threoninum latin |
| PubChem CID | 6288 |
| ChEBI | CHEBI:16857 |
| IUPAC Name | (2 S,3 R)-2-amino-3-hydroxybutanoic acid |
| SMILES | CC(O)C(N)C(O)=O |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity & Availability | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity & Availability | ||||
|
AAJ2229022
|
N/A | 100 g | Plastic Bottle |
Each for $216.78
|
|
||||
|
AAJ22290A1
|
N/A | 1 kg | Plastic Bottle |
Each for $1,541.92
|
|
||||
Chemical Identifiers
| C4H9NO3 | |
| MFCD00064270 | |
| l-threonine, threonine, threonin, h-thr-oh, 2 s,3 r-2-amino-3-hydroxybutanoic acid, s-threonine, l---threonine, 2-amino-3-hydroxybutyric acid, threonine van, threoninum latin | |
| CHEBI:16857 | |
| CC(O)C(N)C(O)=O |
Specifications
| 255°C (decomposition) | |
| C4H9NO3 | |
| MFCD00064270 | |
| l-threonine, threonine, threonin, h-thr-oh, 2 s,3 r-2-amino-3-hydroxybutanoic acid, s-threonine, l---threonine, 2-amino-3-hydroxybutyric acid, threonine van, threoninum latin | |
| CC(O)C(N)C(O)=O | |
| 119.12 | |
| CHEBI:16857 | |
| ≥98.5% | |
| L-Threonine |
| 72-19-5 | |
| CH3CH(OH)CH(NH2)COOH | |
| 1721646 | |
| AYFVYJQAPQTCCC-UHFFFAOYNA-N | |
| (2 S,3 R)-2-amino-3-hydroxybutanoic acid | |
| 6288 | |
| 119.12 | |
| Crystalline Powder |
Safety and Handling
EINECSNumber : 200-774-1
RTECSNumber : XO8590000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only