missing translation for 'onlineSavingsMsg'
Learn More

L-Threonine, ≥98.5%, Ultrapure

Catalog No. AAJ2229022
Change view
Click to view available options
Quantity:
100 g
1 kg
Packaging:
Plastic Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AAJ2229022 100 g Plastic Bottle
AAJ22290A1 1 kg Plastic Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAJ2229022 Supplier Thermo Fisher Scientific Supplier No. J2229022

CAS: 72-19-5 | C4H9NO3 | 119.12 g/mol

Chemical Identifiers

CAS 72-19-5
Molecular Formula C4H9NO3
Molecular Weight (g/mol) 119.12
MDL Number MFCD00064270
InChI Key AYFVYJQAPQTCCC-UHFFFAOYNA-N
Synonym l-threonine, threonine, threonin, h-thr-oh, 2 s,3 r-2-amino-3-hydroxybutanoic acid, s-threonine, l---threonine, 2-amino-3-hydroxybutyric acid, threonine van, threoninum latin
PubChem CID 6288
ChEBI CHEBI:16857
IUPAC Name (2 S,3 R)-2-amino-3-hydroxybutanoic acid
SMILES CC(O)C(N)C(O)=O

Specifications

Melting Point 255°C (decomposition)
Packaging Plastic Bottle
Linear Formula CH3CH(OH)CH(NH2)COOH
Beilstein 1721646
Quantity 100 g
Formula Weight 119.12
Percent Purity ≥98.5%
Physical Form Crystalline Powder
Chemical Name or Material L-Threonine
EINECSNumber 200-774-1
RTECSNumber XO8590000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only