missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals L-Threonine, 98%

CAS: 72-19-5 | C4H9NO3 | 119.12 g/mol

Supplier:  Thermo Scientific Chemicals 138935000

 View more versions of this product

Catalog No. AC138935000


Only null left

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

255.0°C
72-19-5
200ppm max.
Authentic
98%
Glass Bottle
MFCD00064270
300ppm max.
0.1 % max.
Solubility in water: 90g/L (20°C). Other solubilities: insoluble in most common organic solvents
CC(O)C(N)C(O)=O
−28.30 (20.00°C c=6,H2O)
500 g
CHEBI:16857
98%
L-Threonine
Yellow
10ppm max.
0.5 % max. (1 g, 105°C, 3 hrs)
C4H9NO3
CH3CH(OH)CH(NH2)CO2H
− 28.30
15, 9534
l-threonine, threonine, threonin, h-thr-oh, 2s,3r-2-amino-3-hydroxybutanoic acid, s-threonine, l---threonine, 2-amino-3-hydroxybutyric acid, threonine van, threoninum latin
AYFVYJQAPQTCCC-UHFFFAOYNA-N
2-amino-3-hydroxybutanoic acid
119.12
6288
119.12
Crystalline Powder

Chemical Identifiers

72-19-5
119.12
AYFVYJQAPQTCCC-UHFFFAOYNA-N
6288
CC(O)C(N)C(O)=O
C4H9NO3
MFCD00064270
l-threonine, threonine, threonin, h-thr-oh, 2s,3r-2-amino-3-hydroxybutanoic acid, s-threonine, l---threonine, 2-amino-3-hydroxybutyric acid, threonine van, threoninum latin
CHEBI:16857
Product Suggestions

Product Suggestions

Videos
SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only