missing translation for 'onlineSavingsMsg'
Learn More

L-Threitol 98.0+%, TCI America™

Catalog No. T16471G
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T16471G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. T16471G Supplier TCI America Supplier No. T16471G
Only null left

Chemical Identifiers

CAS 2319-57-5
Molecular Formula C4H10O4
Molecular Weight (g/mol) 122.12
MDL Number MFCD00064294
InChI Key UNXHWFMMPAWVPI-IMJSIDKUSA-N
Synonym l-threitol, d-treitol, l-threo-tetritol, 2s,3s-butane-1,2,3,4-tetrol, 2s,3s-butane-1,2,3,4-tetraol, l-1,2,3,4-butanetetrol, l-treitol, l---threitol, 2s,3s-1,2,3,4-butanetetrol
PubChem CID 445969
ChEBI CHEBI:42090
IUPAC Name (2S,3S)-butane-1,2,3,4-tetrol
SMILES C(C(C(CO)O)O)O

Specifications

Melting Point 90°C
Color White
Quantity 1 g
Formula Weight 122.12
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material L-Threitol