missing translation for 'onlineSavingsMsg'
Learn More

L-tert-Leucinol 97.0+%, TCI America™

Catalog No. p-7131849
Change view
missing translation for 'orderingAttributeHoverText'
Quantity:
1 g
5 g
Catalog No. Quantity
L01601G 1 g
L01605G 5 g
2 options

Catalog No. L01601G

Supplier: TCI America L01601G

Only null left

Chemical Identifiers

CAS 112245-13-3
Molecular Formula C6H16NO
Molecular Weight (g/mol) 118.20
MDL Number MFCD00192250
InChI Key JBULSURVMXPBNA-RXMQYKEDSA-O
Synonym l-tert-leucinol, s-tert-leucinol, 2s-2-amino-3,3-dimethylbutan-1-ol, 2s-2-amino-3,3-dimethyl-butan-1-ol, s-2-amino-3,3-dimethylbutanol, s-2-amino-3,3-dimethyl-1-hydroxybutane, r---tert-leucinol, 1-butanol, 2-amino-3,3-dimethyl-, 2s
PubChem CID 2734079
IUPAC Name (2S)-1-hydroxy-3,3-dimethylbutan-2-aminium
SMILES CC(C)(C)[C@H]([NH3+])CO

Specifications

Color White-Yellow
Boiling Point 130°C
Quantity 1 g
Formula Weight 117.19
Percent Purity ≥97.0% (GC,T)
Physical Form Crystalline Lumps
Chemical Name or Material L-tert-Leucinol