missing translation for 'onlineSavingsMsg'
Learn More
Learn More
L-tert-Leucine Methyl Ester 98.0+%, TCI America™
Supplier: TCI America L02551G
Specifications
| L-tert-Leucine Methyl Ester | |
| Colorless | |
| C7H15NO2 | |
| 1 g | |
| H-Tle-OMe, Methyl (S)-2-Amino-3,3-dimethylbutyrate, (S)-2-Amino-3,3-dimethylbutyric Acid Methyl Ester | |
| CC(C)(C)C(C(=O)OC)N | |
| 145.202 | |
| 145.20 | |
| Liquid |
| 63038-26-6 | |
| 69°C | |
| MFCD00137409 | |
| 1993 | |
| WCYLIGGIKNKWQX-RXMQYKEDSA-N | |
| methyl (2S)-2-amino-3,3-dimethylbutanoate | |
| 11309583 | |
| ≥98.0% (GC,T) |
Chemical Identifiers
| 63038-26-6 | |
| 145.202 | |
| WCYLIGGIKNKWQX-RXMQYKEDSA-N | |
| 11309583 | |
| CC(C)(C)C(C(=O)OC)N |
| C7H15NO2 | |
| MFCD00137409 | |
| H-Tle-OMe, Methyl (S)-2-Amino-3,3-dimethylbutyrate, (S)-2-Amino-3,3-dimethylbutyric Acid Methyl Ester | |
| methyl (2S)-2-amino-3,3-dimethylbutanoate |
Safety and Handling
TSCA : No