accessibility menu, dialog, popup

UserName

CAS RN 63038-26-6

CH3H3CCH3OOCH3H2N

CAS RN 63038-26-6

IUPAC Name: methyl 2-amino-3,3-dimethylbutanoate
Synonyms: methyl 2-amino-3,3-dimethylbutanoate
Molecular Weight (g/mol): 145.2
Molecular Formula: C7H15NO2
InChi Key: WCYLIGGIKNKWQX-UHFFFAOYNA-N
SMILES: COC(=O)C(N)C(C)(C)C
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results