missing translation for 'onlineSavingsMsg'
Learn More

L-tert-Leucine Methyl Ester 98.0+%, TCI America™

Catalog No. L0255200MG
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
L0255200MG 200 mg
L02551G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. L0255200MG Supplier TCI America Supplier No. L0255200MG
Only null left

Chemical Identifiers

CAS 63038-26-6
Molecular Formula C7H15NO2
Molecular Weight (g/mol) 145.202
MDL Number MFCD00137409
InChI Key WCYLIGGIKNKWQX-RXMQYKEDSA-N
Synonym H-Tle-OMe, Methyl (S)-2-Amino-3,3-dimethylbutyrate, (S)-2-Amino-3,3-dimethylbutyric Acid Methyl Ester
PubChem CID 11309583
IUPAC Name methyl (2S)-2-amino-3,3-dimethylbutanoate
SMILES CC(C)(C)C(C(=O)OC)N

Specifications

Color Colorless
Boiling Point 69°C
Quantity 200 mg
UN Number 1993
Formula Weight 145.20
Percent Purity ≥98.0% (GC,T)
Physical Form Liquid
Chemical Name or Material L-tert-Leucine Methyl Ester