missing translation for 'onlineSavingsMsg'
Learn More

L-tert-Leucine, 99%, Thermo Scientific™

Chemical Identifiers

CAS 20859-02-3
Molecular Formula C6H13NO2
Molecular Weight (g/mol) 131.175
MDL Number MFCD00064218
InChI Key NPDBDJFLKKQMCM-SCSAIBSYSA-N
Synonym l-tert-leucine, 2s-2-amino-3,3-dimethylbutanoic acid, 3-methyl-l-valine, l-valine,3-methyl, h-tle-oh, s-2-amino-3,3-dimethylbutanoic acid, 3-methyl-l-valin, l-2-amino-3,3-dimethylbutanoic acid, s-2-amino-3,3-dimethylbutyric acid
PubChem CID 164608
IUPAC Name (2S)-2-amino-3,3-dimethylbutanoic acid
SMILES CC(C)(C)C(C(=O)O)N
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC297780050
SDS View Documents
Thermo Scientific Chemicals
297780050
5g Glass bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AC297780250
SDS View Documents
Thermo Scientific Chemicals
297780250
25g Glass bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Chemical Identifiers

20859-02-3
131.175
NPDBDJFLKKQMCM-SCSAIBSYSA-N
164608
CC(C)(C)C(C(=O)O)N
C6H13NO2
MFCD00064218
l-tert-leucine, 2s-2-amino-3,3-dimethylbutanoic acid, 3-methyl-l-valine, l-valine,3-methyl, h-tle-oh, s-2-amino-3,3-dimethylbutanoic acid, 3-methyl-l-valin, l-2-amino-3,3-dimethylbutanoic acid, s-2-amino-3,3-dimethylbutyric acid
(2S)-2-amino-3,3-dimethylbutanoic acid

Specifications

20859-02-3
Authentic
0.2% max. (105°C, 3 hrs)
C6H13NO2
MFCD00064218
0.1% max.
NPDBDJFLKKQMCM-SCSAIBSYSA-N
(2S)-2-amino-3,3-dimethylbutanoic acid
5g
164608
99%
10ppm max.
10ppm max.
99%
Glass bottle
+ 6.30
l-tert-leucine, 2s-2-amino-3,3-dimethylbutanoic acid, 3-methyl-l-valine, l-valine,3-methyl, h-tle-oh, s-2-amino-3,3-dimethylbutanoic acid, 3-methyl-l-valin, l-2-amino-3,3-dimethylbutanoic acid, s-2-amino-3,3-dimethylbutyric acid
CC(C)(C)C(C(=O)O)N
+6.3° (200°C c=4,6N HCl)
131.175
131.17
L-tert-Leucine
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only