missing translation for 'onlineSavingsMsg'
Learn More

L-tert-Leucine, 99%

Catalog No. AC297780050
Change view
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC297780050 5 g Glass Bottle
AC297780250 25 g Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC297780050 Supplier Thermo Scientific Chemicals Supplier No. 297780050
Only null left

CAS: 20859-02-3 | C6H13NO2 | 131.18 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 20859-02-3
Molecular Formula C6H13NO2
Molecular Weight (g/mol) 131.18
MDL Number MFCD00064218
InChI Key NPDBDJFLKKQMCM-SGAVLPGINA-N
Synonym l-tert-leucine, 2s-2-amino-3,3-dimethylbutanoic acid, 3-methyl-l-valine, l-valine,3-methyl, h-tle-oh, s-2-amino-3,3-dimethylbutanoic acid, 3-methyl-l-valin, l-2-amino-3,3-dimethylbutanoic acid, s-2-amino-3,3-dimethylbutyric acid
PubChem CID 164608
IUPAC Name (2S)-2-amino-3,3-dimethylbutanoic acid
SMILES CC(C)(C)[C@H](N)C(O)=O

Specifications

Melting Point >300.0°C
Color White
Heavy Metals (as Pb) 10ppm max.
Infrared Spectrum Authentic
Iron (Fe) 10ppm max.
Loss on Drying 0.2% max. (105°C, 3 hrs)
Assay Percent Range 99%
Packaging Glass Bottle
Specific Rotation + 6.30
Sulfated Ash 0.1% max.
Solubility Information Solubility in water: 125.5g/L (20°C).
Specific Rotation Condition +6.3° (200°C c=4,6N HCl)
Quantity 5 g
Formula Weight 131.17
Percent Purity 99%
Physical Form Powder
Chemical Name or Material L-tert-Leucine
Show More Show Less

RUO – Research Use Only