missing translation for 'onlineSavingsMsg'
Learn More

L-(+)-Tartaric Acid 99.0+%, TCI America™

Catalog No. T0025500G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T0025500G 500 g
T002525G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T0025500G Supplier TCI America Supplier No. T0025500G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 87-69-4
Molecular Formula C4H6O6
Molecular Weight (g/mol) 150.09
MDL Number MFCD00064207
InChI Key FEWJPZIEWOKRBE-UHFFFAOYNA-N
Synonym l-tartaric acid, l-+-tartaric acid, l +-tartaric acid, 2r,3r-2,3-dihydroxysuccinic acid, tartaric acid, 2r,3r-2,3-dihydroxybutanedioic acid, r,r-tartaric acid, #NAME?, dextrotartaric acid, l-threaric acid
PubChem CID 444305
ChEBI CHEBI:15671
IUPAC Name 2,3-dihydroxybutanedioic acid
SMILES OC(C(O)C(O)=O)C(O)=O

Specifications

Color White
Quantity 500 g
Physical Form Crystalline Powder
Formula Weight 150.09
Percent Purity ≥99.0% (T)
Chemical Name or Material L-(+)-Tartaric Acid
EINECSNumber (2)-1456
RTECSNumber WW7875000
TSCA Yes