missing translation for 'onlineSavingsMsg'
Learn More

L-(+)-Tartaric Acid (Granular/Certified ACS), Fisher Chemical™

Catalog No. p-194571
Change view
Click to view available options
Quantity:
500 g
3 kg
Packaging:
Poly Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
A314500 500 g Poly Bottle
A3143 3 kg Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A314500 Supplier Thermo Fisher Scientific Supplier No. FLA314500
Only null left

Chemical Identifiers

CAS 87-69-4
Molecular Formula C4H6O6
Molecular Weight (g/mol) 150.09
MDL Number MFCD00064207
InChI Key FEWJPZIEWOKRBE-UHFFFAOYNA-N
Synonym l-tartaric acid, l-+-tartaric acid, l +-tartaric acid, 2r,3r-2,3-dihydroxysuccinic acid, tartaric acid, 2r,3r-2,3-dihydroxybutanedioic acid, r,r-tartaric acid, +-l-tartaric acid, dextrotartaric acid, l-threaric acid
PubChem CID 444305
ChEBI CHEBI:15671
IUPAC Name 2,3-dihydroxybutanedioic acid
SMILES OC(C(O)C(O)=O)C(O)=O

Specifications

pH 1.6
Melting Point 168°C
Color White
Assay Percent Range ≥99.0 %
Linear Formula HOOC(CHOH)2COOH
Formula Weight 150.09
Percent Purity ≥99.0%
Grade Certified ACS
Density 1.76g/mL
Quantity 500 g
Ignition Residue 0.02% max.
Packaging Poly Bottle
Physical Form Solid
Vapor Pressure <0.1 mbar at 20°C
Chemical Name or Material L-(+)-Tartaric Acid
Show More Show Less