missing translation for 'onlineSavingsMsg'
Learn More

L(+)-Tartaric acid, EMSURE™ MilliporeSigma™

Catalog No. M1008040250
Change view
Click to view available options
Quantity:
250 g
1 kg
Packaging:
Plastic Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
M1008040250 250 g Plastic Bottle
M1008041000 1 kg Plastic Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M1008040250 Supplier Technidata Supplier No. 1.00804.0250

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 87-69-4
Molecular Formula C4H6O6
Molecular Weight (g/mol) 150.09
MDL Number MFCD00064207
InChI Key FEWJPZIEWOKRBE-UHFFFAOYNA-N
Synonym l-tartaric acid, l-+-tartaric acid, l +-tartaric acid, 2r,3r-2,3-dihydroxysuccinic acid, tartaric acid, 2r,3r-2,3-dihydroxybutanedioic acid, r,r-tartaric acid, +-l-tartaric acid, dextrotartaric acid, l-threaric acid
PubChem CID 444305
ChEBI CHEBI:15671
IUPAC Name 2,3-dihydroxybutanedioic acid
SMILES OC(C(O)C(O)=O)C(O)=O

Specifications

pH 1.6 (100g/L, H2O, 25°C)
Melting Point 168°C to 170°C
Color White
Density 1.76 g/3 (20 °C)
Quantity 250 g
Flash Point 210°C
Packaging Plastic Bottle
Physical Form Solid
Solubility Information 1390g/L
Formula Weight 150.08
Percent Purity >99.5%
Grade ACS
Show More Show Less
EINECSNumber 201-766-0