missing translation for 'onlineSavingsMsg'
Learn More
Learn More
L(+)-Tartaric acid, 99+%
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Melting Point | 168.0°C to 172.0°C |
| Color | White or Colorless |
| Assay Percent Range | 99+% |
| Linear Formula | HO2CCH(OH)CH(OH)CO2H |
| Solubility Information | Solubility in water: 1390g/L (20°C). Other solubilities: soluble in methanol, ethanol, propanol and, glycerol, 4g/L ether, insoluble in chloroform |
| IUPAC Name | 2,3-dihydroxybutanedioic acid |
| Formula Weight | 150.09 |
| Percent Purity | ≥99% |
| Quantity | 500 g |
| Flash Point | 210°C |
| Show More |
Safety and Handling
| Hazard Category | Danger |
RUO – Research Use Only