Learn More
L-(+)-Tartaric acid, 99%
CAS: 87-69-4 | C4H6O6 | 150.09 g/mol
Supplier: Thermo Scientific Chemicals A1366822
Description
L-(+)-Tartaric acid is widely utilized in pharmaceutical industries. It is used in soft drinks, confectionaries, food products, gelatin desserts and as a buffering agent. It forms a compound, TiCl2(O-i-Pr)2 with Diels-Alder catalyst and acta as a chelate agent in metal industries. Owing to its efficient chelating property towards metal ions, it is used in farming and metal industries for complexing micronutrients and for cleaning metal surfaces, respectively.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsL-(+)-Tartaric acid is widely utilized in pharmaceutical industries. It is used in soft drinks, confectionaries, food products, gelatin desserts and as a buffering agent. It forms a compound, TiCl2(O-i-Pr)2 with Diels-Alder catalyst and acta as a chelate agent in metal industries. Owing to its efficient chelating property towards metal ions, it is used in farming and metal industries for complexing micronutrients and for cleaning metal surfaces, respectively.
Solubility
Soluble in water, methanol and acetone.
Notes
Incompatible with oxidizing agents, bases and reducing agents.
Specifications
| L-(+)-Tartaric acid | |
| 168°C to 170°C | |
| C4H6O6 | |
| 1725147 | |
| l-tartaric acid, l-+-tartaric acid, l +-tartaric acid, 2r,3r-2,3-dihydroxysuccinic acid, tartaric acid, 2r,3r-2,3-dihydroxybutanedioic acid, r,r-tartaric acid, #NAME?, dextrotartaric acid, l-threaric acid | |
| FEWJPZIEWOKRBE-UHFFFAOYNA-N | |
| (2R,3R)-2,3-dihydroxybutanedioic acid | |
| 444305 | |
| 150.09 | |
| 1.76 | |
| Odorless |
| 2.1 | |
| 87-69-4 | |
| MFCD00064207 | |
| 149070 | |
| Soluble in water,methanol and acetone. | |
| OC(C(O)C(O)=O)C(O)=O | |
| 150.09 | |
| CHEBI:15671 | |
| 99% | |
| 100 g | |
| +12.4° (c=20 in Water) |
Chemical Identifiers
| 87-69-4 | |
| 150.09 | |
| FEWJPZIEWOKRBE-UHFFFAOYNA-N | |
| 444305 | |
| (2R,3R)-2,3-dihydroxybutanedioic acid |
| C4H6O6 | |
| MFCD00064207 | |
| l-tartaric acid, l-+-tartaric acid, l +-tartaric acid, 2r,3r-2,3-dihydroxysuccinic acid, tartaric acid, 2r,3r-2,3-dihydroxybutanedioic acid, r,r-tartaric acid, #NAME?, dextrotartaric acid, l-threaric acid | |
| CHEBI:15671 | |
| OC(C(O)C(O)=O)C(O)=O |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P280-P305+P351+P338-P310
H318-H500
EINECSNumber : 201-766-0
RTECSNumber : WW7875000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only