missing translation for 'onlineSavingsMsg'
Learn More

L-(+)-Tartaric acid, 99%

Catalog No. AAA1366836
Change view
Click to view available options
Quantity:
100 g
500 g
2500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1366836 500 g
AAA1366822 100 g
AAA136680E 2500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1366836 Supplier Thermo Scientific Chemicals Supplier No. A1366836
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 87-69-4 | C4H6O6 | 150.09 g/mol

L-(+)-Tartaric acid is widely utilized in pharmaceutical industries. It is used in soft drinks, confectionaries, food products, gelatin desserts and as a buffering agent. It forms a compound, TiCl2(O-i-Pr)2 with Diels-Alder catalyst and acta as a chelate agent in metal industries. Owing to its efficient chelating property towards metal ions, it is used in farming and metal industries for complexing micronutrients and for cleaning metal surfaces, respectively.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
L-(+)-Tartaric acid is widely utilized in pharmaceutical industries. It is used in soft drinks, confectionaries, food products, gelatin desserts and as a buffering agent. It forms a compound, TiCl2(O-i-Pr)2 with Diels-Alder catalyst and acta as a chelate agent in metal industries. Owing to its efficient chelating property towards metal ions, it is used in farming and metal industries for complexing micronutrients and for cleaning metal surfaces, respectively.

Solubility
Soluble in water, methanol and acetone.

Notes
Incompatible with oxidizing agents, bases and reducing agents.

Chemical Identifiers

CAS 87-69-4
Molecular Formula C4H6O6
Molecular Weight (g/mol) 150.09
MDL Number MFCD00064207
InChI Key FEWJPZIEWOKRBE-UHFFFAOYNA-N
Synonym l-tartaric acid, l-+-tartaric acid, l +-tartaric acid, 2r,3r-2,3-dihydroxysuccinic acid, tartaric acid, 2r,3r-2,3-dihydroxybutanedioic acid, r,r-tartaric acid, #NAME?, dextrotartaric acid, l-threaric acid
PubChem CID 444305
ChEBI CHEBI:15671
IUPAC Name (2R,3R)-2,3-dihydroxybutanedioic acid
SMILES OC(C(O)C(O)=O)C(O)=O

Specifications

pH 2.1
Melting Point 168°C to 170°C
Density 1.76
Quantity 500 g
Odor Odorless
Optical Rotation +12.4° (c=20 in Water)
Beilstein 1725147
Merck Index 149070
Solubility Information Soluble in water,methanol and acetone.
Formula Weight 150.09
Percent Purity 99%
Chemical Name or Material L-(+)-Tartaric acid
Show More Show Less
Hazard Category H318-H500
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P280-P305+P351+P338-P310
EINECSNumber 201-766-0
RTECSNumber WW7875000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only