missing translation for 'onlineSavingsMsg'
Learn More

L-(-)-Talose 98.0+%, TCI America™

Catalog No. T1767100MG
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T1767100MG 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. T1767100MG Supplier TCI America Supplier No. T1767100MG
Only null left

Chemical Identifiers

CAS 23567-25-1
Molecular Formula C6H12O6
Molecular Weight (g/mol) 180.156
MDL Number MFCD00135850
InChI Key GZCGUPFRVQAUEE-OMMKOOBNSA-N
Synonym l-talose, l---talose, 2r,3r,4r,5s-2,3,4,5,6-pentahydroxyhexanal, aldehydo-l-talose
PubChem CID 90173
ChEBI CHEBI:86058
IUPAC Name (2R,3R,4R,5S)-2,3,4,5,6-pentahydroxyhexanal
SMILES C(C(C(C(C(C=O)O)O)O)O)O

Specifications

Melting Point 133°C
Color White
Quantity 100 mg
Formula Weight 180.16
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material L-(-)-Talose