missing translation for 'onlineSavingsMsg'
Learn More

L-Talitol 98.0+%, TCI America™

Catalog No. T2536100MG
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T2536100MG 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. T2536100MG Supplier TCI America Supplier No. T2536100MG
Only null left

Chemical Identifiers

CAS 60660-58-4
Molecular Formula C6H14O6
Molecular Weight (g/mol) 182.172
InChI Key FBPFZTCFMRRESA-OMMKOOBNSA-N
PubChem CID 1549089
IUPAC Name (2S,3S,4R,5S)-hexane-1,2,3,4,5,6-hexol
SMILES C(C(C(C(C(CO)O)O)O)O)O

Specifications

Color White
Quantity 100 mg
Formula Weight 182.17
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material L-Talitol