missing translation for 'onlineSavingsMsg'
Learn More
Learn More
L(-)-Sorbose, 98%
CAS: 87-79-6 | C6H12O6 | 180.16 g/mol
Supplier: Thermo Scientific Chemicals 225960010
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
| L(-)-Sorbose | |
| 87-79-6 | |
| White | |
| Authentic | |
| Plastic bottle | |
| MFCD00151097 | |
| 15,8852 | |
| -43 | |
| Solubility in water: soluble. Other solubilities: almost insoluble in alcohol,slightly soluble in ethanol,ethylether,methanol | |
| C(C(C(C(C(=O)CO)O)O)O)O | |
| 180.16 | |
| CHEBI:13172 | |
| 98% |
| 99% | |
| 158°C to 160°C | |
| 1% max. | |
| 97.5% min. (ELSD) (HPLC) | |
| C6H12O6 | |
| 1 kg | |
| - 43.00 (20.00°C c=5,H2O,23 C) | |
| l---sorbose, 3s,4r,5s-1,3,4,5,6-pentahydroxyhexan-2-one, sorbinose, keto-l-sorbose, esorben, sorbin, l--sorbose, hsdb 780, l-sorbinose, l-xylo-2-hexulose | |
| BJHIKXHVCXFQLS-OTWZMJIISA-N | |
| (3S,4R,5S)-1,3,4,5,6-pentahydroxyhexan-2-one | |
| 6904 | |
| 180.16 | |
| Crystalline Powder, Crystals or Chunks |
Chemical Identifiers
| 87-79-6 | |
| BJHIKXHVCXFQLS-OTWZMJIISA-N | |
| 6904 | |
| (3S,4R,5S)-1,3,4,5,6-pentahydroxyhexan-2-one |
| MFCD00151097 | |
| l---sorbose, 3s,4r,5s-1,3,4,5,6-pentahydroxyhexan-2-one, sorbinose, keto-l-sorbose, esorben, sorbin, l--sorbose, hsdb 780, l-sorbinose, l-xylo-2-hexulose | |
| CHEBI:13172 | |
| C(C(C(C(C(=O)CO)O)O)O)O |
Safety and Handling
EINECSNumber : 201-771-8
RTECSNumber : WG3195025
TSCA : TSCA
RUO – Research Use Only