accessibility menu, dialog, popup

UserName

CAS RN 87-79-6

IUPAC Name: 1,3,4,5,6-pentahydroxyhexan-2-one
Synonyms: sorbose
Molecular Weight (g/mol): 180.16
Molecular Formula: C6H12O6
InChi Key: BJHIKXHVCXFQLS-UHFFFAOYNA-N
SMILES: OCC(O)C(O)C(O)C(=O)CO
Molecular Weight (g/mol) 
  • (4)
  • (3)
Quantity 
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
Percent Purity 
  • (2)
  • (1)
  • (3)
Physical Form 
  • (3)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (1)
  • (3)
  • (4)
  • (3)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (3)
  • (3)
  • (3)