missing translation for 'onlineSavingsMsg'
Learn More

L(-)-Sorbose, 98%

Catalog No. AC225961000
Change view
Click to view available options
Quantity:
100 g
250 g
1 kg
Packaging:
Plastic bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC225961000 100 g Plastic bottle
AC225962500 250 g Plastic bottle
AC225960010 1 kg Plastic bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC225961000 Supplier Thermo Scientific Chemicals Supplier No. 225961000

CAS: 87-79-6 | C6H12O6 | 180.16 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 87-79-6
MDL Number MFCD00151097
InChI Key BJHIKXHVCXFQLS-OTWZMJIISA-N
Synonym l---sorbose, 3s,4r,5s-1,3,4,5,6-pentahydroxyhexan-2-one, sorbinose, keto-l-sorbose, esorben, sorbin, l--sorbose, hsdb 780, l-sorbinose, l-xylo-2-hexulose
PubChem CID 6904
ChEBI CHEBI:13172
IUPAC Name (3S,4R,5S)-1,3,4,5,6-pentahydroxyhexan-2-one
SMILES C(C(C(C(C(=O)CO)O)O)O)O

Specifications

Melting Point 158°C to 160°C
Color White
Loss on Drying 1% max.
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (ELSD) (HPLC)
Packaging Plastic bottle
Molecular Formula C6H12O6
Quantity 100 g
Merck Index 15,8852
Specific Rotation Condition - 43.00 (20.00°C c=5,H2O,23 C)
Specific Rotation -43
Solubility Information Solubility in water: soluble. Other solubilities: almost insoluble in alcohol,slightly soluble in ethanol,ethylether,methanol
Molecular Weight (g/mol) 180.16
Formula Weight 180.16
Percent Purity 98%
Physical Form Crystalline Powder, Crystals or Chunks
Chemical Name or Material L(-)-Sorbose, 99%
Show More Show Less
EINECSNumber 201-771-8
RTECSNumber WG3195025
TSCA TSCA

RUO – Research Use Only