missing translation for 'onlineSavingsMsg'
Learn More

L-(-)-Sorbose, ≥98%, For HPLC analysis, MP Biomedicals™

Catalog No. p-4404535
Change view
missing translation for 'orderingAttributeHoverText'
Quantity:
1 kg
500 g
Catalog No. Quantity
ICN10293990 500 g
ICN10293991 1 kg
2 options

Catalog No. ICN10293990

Supplier: MP Biomedicals Inc 0210293990

Only null left

Chemical Identifiers

CAS 87-79-6
Molecular Formula C6H12O6
Molecular Weight (g/mol) 180.156
InChI Key BJHIKXHVCXFQLS-OTWZMJIISA-N
Synonym l---sorbose, 3s,4r,5s-1,3,4,5,6-pentahydroxyhexan-2-one, sorbinose, keto-l-sorbose, esorben, sorbin, l--sorbose, hsdb 780, l-sorbinose, l-xylo-2-hexulose
PubChem CID 6904
ChEBI CHEBI:13172
IUPAC Name (3S,4R,5S)-1,3,4,5,6-pentahydroxyhexan-2-one
SMILES C(C(C(C(C(=O)CO)O)O)O)O

Specifications

Color White to Yellow
Quantity 500 g
Formula Weight 180.2
Percent Purity ≥98%
Physical Form Crystalline Powder
Chemical Name or Material L-(-)-Sorbose, For HPLC analysis