missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ L-Serine methyleester hydrochloride, 98%

$64.50 - $213.41

Chemical Identifiers

CAS 5680-80-8
Molecular Formula C4H10ClNO3
Molecular Weight (g/mol) 155.578
MDL Number MFCD00063680
InChI Key NDBQJIBNNUJNHA-DFWYDOINSA-N
Synonym l-serine methyl ester hydrochloride, h-ser-ome.hcl, methyl l-serinate hydrochloride, s-methyl 2-amino-3-hydroxypropanoate hydrochloride, methyl 2s-2-amino-3-hydroxypropanoate hydrochloride, l-serine, methyl ester, hydrochloride, l-methyl serinate hcl, serine methyl ester hydrochloride, l-serine methyl ester hcl, h-ser-ome-hcl
PubChem CID 2723730
IUPAC Name methyl (2S)-2-amino-3-hydroxypropanoate;hydrochloride
SMILES COC(=O)C(CO)N.Cl
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Packaging Price Quantity & Availability  
AC132670050
View Documents Promotion Details
Thermo Scientific™
132670050
5g Glass bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AC132670250
View Documents Promotion Details
Thermo Scientific™
132670250
25g Glass bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Chemical Identifiers

5680-80-8
155.578
NDBQJIBNNUJNHA-DFWYDOINSA-N
2723730
COC(=O)C(CO)N.Cl
C4H10ClNO3
MFCD00063680
l-serine methyl ester hydrochloride, h-ser-ome.hcl, methyl l-serinate hydrochloride, s-methyl 2-amino-3-hydroxypropanoate hydrochloride, methyl 2s-2-amino-3-hydroxypropanoate hydrochloride, l-serine, methyl ester, hydrochloride, l-methyl serinate hcl, serine methyl ester hydrochloride, l-serine methyl ester hcl, h-ser-ome-hcl
methyl (2S)-2-amino-3-hydroxypropanoate;hydrochloride

Specifications

5680-80-8
98%
HOCH2CH(NH2)CO2CH3·HCl
+ 4.50
l-serine methyl ester hydrochloride, h-ser-ome.hcl, methyl l-serinate hydrochloride, s-methyl 2-amino-3-hydroxypropanoate hydrochloride, methyl 2s-2-amino-3-hydroxypropanoate hydrochloride, l-serine, methyl ester, hydrochloride, l-methyl serinate hcl, serine methyl ester hydrochloride, l-serine methyl ester hcl, h-ser-ome-hcl
1% Max. (K.F.)
COC(=O)C(CO)N.Cl
methyl (2S)-2-amino-3-hydroxypropanoate;hydrochloride
2723730
98%
Authentic
C4H10ClNO3
MFCD00063680
04, 506
0.1% max.
NDBQJIBNNUJNHA-DFWYDOINSA-N
+ 4.50 (20.00°C c=2,methanol)
155.578
155.58
L-Serine methyl ester hydrochloride
Videos
SDS
Documents

Documents

RUO – Research Use Only