missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals L-Serine methyl ester hydrochloride, 98%

Catalog No. AC132670250
Change view
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC132670250 25 g Glass Bottle
AC132670050 5 g Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC132670250 Supplier Thermo Scientific Chemicals Supplier No. 132670250
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 5680-80-8 | C4H10ClNO3 | 155.578 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 5680-80-8
Molecular Formula C4H10ClNO3
Molecular Weight (g/mol) 155.578
MDL Number MFCD00063680
InChI Key NDBQJIBNNUJNHA-DFWYDOINSA-N
Synonym l-serine methyl ester hydrochloride, h-ser-ome.hcl, methyl l-serinate hydrochloride, s-methyl 2-amino-3-hydroxypropanoate hydrochloride, methyl 2s-2-amino-3-hydroxypropanoate hydrochloride, l-serine, methyl ester, hydrochloride, l-methyl serinate hcl, serine methyl ester hydrochloride, l-serine methyl ester hcl, h-ser-ome-hcl
PubChem CID 2723730
IUPAC Name methyl (2S)-2-amino-3-hydroxypropanoate;hydrochloride
SMILES COC(=O)C(CO)N.Cl

Specifications

Melting Point 160.0°C to 165.0°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass Bottle
Linear Formula HOCH2CH(NH2)CO2CH3·HCl
Specific Rotation + 4.50
Beilstein 04, 506
Sulfated Ash 0.1% max.
Solubility Information Solubility in water: soluble. Other solubilities: soluble in methanol
Water 1% Max. (K.F.)
Specific Rotation Condition + 4.50 (20.00°C c=2,methanol)
Quantity 25 g
Formula Weight 155.58
Percent Purity 98%
Physical Form Crystalline Powder or Crystals
Chemical Name or Material L-Serine methyl ester hydrochloride
Show More Show Less

RUO – Research Use Only