missing translation for 'onlineSavingsMsg'
Learn More

L-Serine, FCC, 98.5-101.5%, Spectrum™ Chemical

Catalog No. p-7157839
Change view
Click to view available options
Quantity:
25 g
100 g
1 kg
Packaging:
Poly Bottle
Poly Pail
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18603979 25 g Poly Bottle
18603980 100 g Poly Bottle
18603981 1 kg Poly Pail
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 18603979 Supplier Spectrum Chemical Mfg Cor Supplier No. S102125GM
Only null left

S1021, 56-45-1, C3H7NO3

Spectrum™ Chemical L-Serine, FCC - The FCC grade meets the requirements of the Food Chemical Codex indicates and is suitable for all food, beverage and nutritional supplement applications. Spectrum Chemical offers over 300 Food grade chemical ingredients packaged in laboratory size bottles to production drum quantities and are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 56-45-1
Molecular Formula C3H7NO3
Molecular Weight (g/mol) 105.09
InChI Key MTCFGRXMJLQNBG-UHFFFAOYNA-N
IUPAC Name 2-amino-3-hydroxypropanoic acid
SMILES NC(CO)C(O)=O

Specifications

CAS Max % 100%
Ignition Residue 0.1%
Loss on Drying 0.3%
Packaging Poly Bottle
Quantity 25 g
Optical Rotation +13.6° to +160°
Percent Purity 98.5 to 101.5%
Grade FCC