missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MilliporeSigma™ L-Serine, Calbiochem™,
Supplier: MilliporeSigma™ 5650100GM
Specifications
| 56-45-1 | |
| C3H7NO3 | |
| l-serine, serine, s-2-amino-3-hydroxypropanoic acid, s-serine, beta-hydroxyalanine, h-ser-oh, l-ser, l---serine, serine, l, 2s-2-amino-3-hydroxypropanoic acid | |
| C(C(C(=O)O)N)O | |
| 100 g | |
| 5951 | |
| ≥99% (Assay (Titration)) |
| White | |
| Glass Bottle | |
| MTCFGRXMJLQNBG-REOHCLBHSA-N | |
| (2S)-2-amino-3-hydroxypropanoic acid | |
| 105.093 | |
| CHEBI:17115 | |
| Solid |
Chemical Identifiers
| 56-45-1 | |
| 105.093 | |
| l-serine, serine, s-2-amino-3-hydroxypropanoic acid, s-serine, beta-hydroxyalanine, h-ser-oh, l-ser, l---serine, serine, l, 2s-2-amino-3-hydroxypropanoic acid | |
| CHEBI:17115 | |
| C(C(C(=O)O)N)O |
| C3H7NO3 | |
| MTCFGRXMJLQNBG-REOHCLBHSA-N | |
| 5951 | |
| (2S)-2-amino-3-hydroxypropanoic acid |