missing translation for 'onlineSavingsMsg'
Learn More
Learn More
L-Serine benzyl ester hydrochloride, 98%
CAS: 60022-62-0 | C10H14ClNO3 | 231.68 g/mol
$86.14 - $714.95
Chemical Identifiers
| CAS | 60022-62-0 |
|---|---|
| Molecular Formula | C10H14ClNO3 |
| Molecular Weight (g/mol) | 231.68 |
| MDL Number | MFCD00038955 |
| InChI Key | MGZWCDQAKCHOBX-UHFFFAOYNA-N |
| Synonym | l-serine benzyl ester hydrochloride, h-ser-obzl.hcl, ser bzl .hcl, h-ser.obzl.hcl, benzyl 2s-2-amino-3-hydroxypropanoate hydrochloride, l-serine, phenylmethyl ester, hydrochloride, l-serinebenzylesterhydrochloride, h-ser-obzl. hcl, pubchem19043, h-ser-obn hydrochloride |
| PubChem CID | 12930233 |
| SMILES | Cl.NC(CO)C(=O)OCC1=CC=CC=C1 |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAH2879003
|
Thermo Scientific Chemicals
H2879003 |
1 g |
Each for $86.14
|
|
|||||
|
AAH2879006
|
Thermo Scientific Chemicals
H2879006 |
5 g |
Each for $247.14
|
|
|||||
|
AAH2879014
|
Thermo Scientific Chemicals
H2879014 |
25 g |
Each for $714.95
|
|
|||||
Description
It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsIt is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff.
Solubility
Soluble in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
Chemical Identifiers
| 60022-62-0 | |
| 231.68 | |
| MGZWCDQAKCHOBX-UHFFFAOYNA-N | |
| 12930233 |
| C10H14ClNO3 | |
| MFCD00038955 | |
| l-serine benzyl ester hydrochloride, h-ser-obzl.hcl, ser bzl .hcl, h-ser.obzl.hcl, benzyl 2s-2-amino-3-hydroxypropanoate hydrochloride, l-serine, phenylmethyl ester, hydrochloride, l-serinebenzylesterhydrochloride, h-ser-obzl. hcl, pubchem19043, h-ser-obn hydrochloride | |
| Cl.NC(CO)C(=O)OCC1=CC=CC=C1 |
Specifications
| ∽175°C (decomposition) | |
| C10H14ClNO3 | |
| l-serine benzyl ester hydrochloride, h-ser-obzl.hcl, ser bzl .hcl, h-ser.obzl.hcl, benzyl 2s-2-amino-3-hydroxypropanoate hydrochloride, l-serine, phenylmethyl ester, hydrochloride, l-serinebenzylesterhydrochloride, h-ser-obzl. hcl, pubchem19043, h-ser-obn hydrochloride | |
| MGZWCDQAKCHOBX-UHFFFAOYNA-N | |
| benzyl (2S)-2-amino-3-hydroxypropanoate;hydrochloride | |
| 231.68 | |
| −12° (c=4 in Water) | |
| 98% |
| 60022-62-0 | |
| MFCD00038955 | |
| Soluble in water. | |
| Cl.NC(CO)C(=O)OCC1=CC=CC=C1 | |
| 1 g | |
| 12930233 | |
| 231.68 | |
| L-Serine benzyl ester hydrochloride |
Safety and Handling
TSCA : No
Recommended Storage : Keep cold
RUO – Research Use Only