missing translation for 'onlineSavingsMsg'
Learn More

L-Serine benzyl ester hydrochloride, 98%

CAS: 60022-62-0 | C10H14ClNO3 | 231.68 g/mol

Supplier:  Thermo Scientific Chemicals H2879014

 View more versions of this product

Catalog No. AAH2879014


Only null left
Explore available promotions

Description

Description

It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff.

Solubility
Soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
Specifications

Specifications

∽175°C (decomposition)
60022-62-0
MFCD00038955
Soluble in water.
Cl.NC(CO)C(=O)OCC1=CC=CC=C1
25 g
12930233
231.68
L-Serine benzyl ester hydrochloride
C10H14ClNO3
l-serine benzyl ester hydrochloride, h-ser-obzl.hcl, ser bzl .hcl, h-ser.obzl.hcl, benzyl 2s-2-amino-3-hydroxypropanoate hydrochloride, l-serine, phenylmethyl ester, hydrochloride, l-serinebenzylesterhydrochloride, h-ser-obzl. hcl, pubchem19043, h-ser-obn hydrochloride
MGZWCDQAKCHOBX-UHFFFAOYNA-N
benzyl (2S)-2-amino-3-hydroxypropanoate;hydrochloride
231.68
−12° (c=4 in Water)
98%

Chemical Identifiers

60022-62-0
231.68
MGZWCDQAKCHOBX-UHFFFAOYNA-N
12930233
C10H14ClNO3
MFCD00038955
l-serine benzyl ester hydrochloride, h-ser-obzl.hcl, ser bzl .hcl, h-ser.obzl.hcl, benzyl 2s-2-amino-3-hydroxypropanoate hydrochloride, l-serine, phenylmethyl ester, hydrochloride, l-serinebenzylesterhydrochloride, h-ser-obzl. hcl, pubchem19043, h-ser-obn hydrochloride
Cl.NC(CO)C(=O)OCC1=CC=CC=C1
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

TSCA : No

Recommended Storage : Keep cold

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only