missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific Chemicals L-Serine, 99%
CAS: 56-45-1 | C3H7NO3 | 105.093 g/mol
Supplier: Thermo Scientific Chemicals 132661000
| Quantity | 100 g |
|---|---|
| Packaging | Glass Bottle |
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 56-45-1 | |
| 105.093 | |
| l-serine, serine, s-2-amino-3-hydroxypropanoic acid, s-serine, beta-hydroxyalanine, h-ser-oh, l-ser, l---serine, serine, l, 2s-2-amino-3-hydroxypropanoic acid | |
| CHEBI:17115 | |
| C(C(C(=O)O)N)O |
| C3H7NO3 | |
| MTCFGRXMJLQNBG-REOHCLBHSA-N | |
| 5951 | |
| (2S)-2-amino-3-hydroxypropanoic acid |
Specifications
| L-Serine | |
| 200ppm max. | |
| 1ppm max. | |
| White | |
| 10ppm max. | |
| 10ppm max. | |
| 0.2% (105°C, 3 hrs) max. | |
| Glass Bottle | |
| HOCH2CH(NH2)CO2H | |
| 200ppm max. | |
| l-serine, serine, s-2-amino-3-hydroxypropanoic acid, s-serine, beta-hydroxyalanine, h-ser-oh, l-ser, l---serine, serine, l, 2s-2-amino-3-hydroxypropanoic acid | |
| Solubility in water: 250g/L (20°C). Other solubilities: insoluble in alcohol, ether and benzene, soluble in mineral acids, insoluble in organic solvents, insoluble in ethanol and diethylether | |
| MTCFGRXMJLQNBG-REOHCLBHSA-N | |
| 1.6000g/mL | |
| (2S)-2-amino-3-hydroxypropanoic acid | |
| 105.093 | |
| CHEBI:17115 | |
| 99% | |
| Crystalline Powder, Crystals or Crystalline Needles |
| 222.0°C | |
| 56-45-1 | |
| 5.0 to 6.5 (2.5% soln. at 25°C) | |
| 200ppm max. | |
| Authentic | |
| 99% | |
| 0.5% (TLC) max. | |
| C3H7NO3 | |
| 15.2 | |
| 95% (c=10, H2O, 430nm) 1cm cellmin. | |
| 0.1% max. | |
| 0150.0°C | |
| C(C(C(=O)O)N)O | |
| 15.20 (20.00°C c=10,2N HCl) | |
| 100 g | |
| 5951 | |
| 105.09 | |
| 1.6 |