missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ L-Serine, 99%
Supplier: Thermo Scientific™ 132660025
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
| L-Serine | |
| 200ppm max. | |
| 200ppm max. | |
| Authentic | |
| 99% | |
| 0.5% (TLC) max. | |
| Plastic Bottle | |
| 15.2 | |
| 95% (c=10, H2O, 430nm) 1cm cellmin. | |
| 0.1% max. | |
| MTCFGRXMJLQNBG-REOHCLBHSA-N | |
| 1.6000g/mL | |
| (2S)-2-amino-3-hydroxypropanoic acid | |
| 2.5kg | |
| CHEBI:17115 | |
| 99% |
| 56-45-1 | |
| 1ppm max. | |
| 10ppm max. | |
| 10ppm max. | |
| 0.2% (105°C, 3 hrs) max. | |
| C3H7NO3 | |
| HOCH2CH(NH2)CO2H | |
| 200ppm max. | |
| l-serine, serine, s-2-amino-3-hydroxypropanoic acid, s-serine, beta-hydroxyalanine, h-ser-oh, l-ser, l---serine, serine, l, 2s-2-amino-3-hydroxypropanoic acid | |
| 0150.0°C | |
| C(C(C(=O)O)N)O | |
| 15.20 (20.00°C c=10,2N HCl) | |
| 105.093 | |
| 5951 | |
| 105.09 | |
| 1.6 |