missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals L-Serine, 99%

Catalog No. AAA1117922
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1117922 100 g
AAA1117914 25 g
AAA1117936 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1117922 Supplier Thermo Scientific Chemicals Supplier No. A1117922
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 56-45-1 | C3H7NO3 | 105.093 g/mol

L-serine is used as a dietary supplement to improve health under certain conditions of malnutrition or physical stress, as a cosmetic incredient, as ingredients for cell culture media. It is used as a precursor to several amino acids, including glycine, cysteine and other metabolites such as sphingolipids and folate. It is used in bio-synthesis of purines and pyrimidines. It plays a vital role in catalytic function of enzymes. Further, L-serine and its metabolic products are essential for cell proliferation and specific functions of the central nervous system. .

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
L-serine is used as a dietary supplement to improve health under certain conditions of malnutrition or physical stress, as a cosmetic incredient, as ingredients for cell culture media. It is used as a precursor to several amino acids, including glycine, cysteine and other metabolites such as sphingolipids and folate. It is used in bio-synthesis of purines and pyrimidines. It plays a vital role in catalytic function of enzymes. Further, L-serine and its metabolic products are essential for cell proliferation and specific functions of the central nervous system. .

Solubility
Soluble in water. Insoluble in ethanol, ether and benzene.

Notes
Store in a cool place. Incompatible with strong oxidizing agents.

Chemical Identifiers

CAS 56-45-1
Molecular Formula C3H7NO3
Molecular Weight (g/mol) 105.093
MDL Number MFCD00064224
InChI Key MTCFGRXMJLQNBG-REOHCLBHSA-N
Synonym l-serine, serine, s-2-amino-3-hydroxypropanoic acid, s-serine, beta-hydroxyalanine, h-ser-oh, l-ser, l---serine, serine, l, 2s-2-amino-3-hydroxypropanoic acid
PubChem CID 5951
ChEBI CHEBI:17115
IUPAC Name (2S)-2-amino-3-hydroxypropanoic acid
SMILES C(C(C(=O)O)N)O

Specifications

Melting Point 214°C to 224°C (decomposition)
Beilstein 1721404
Merck Index 14,8460
Solubility Information Soluble in water. Insoluble in ethanol,ether and benzene.
Quantity 100 g
Formula Weight 105.09
Optical Rotation +15° (c=9 in 1N HCl)
Percent Purity 99%
Chemical Name or Material L-Serine
EINECSNumber 200-274-3
RTECSNumber VT8100000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only